Learn More
Thermo Scientific™ ART™ Barrier Pipette Tips in Lift-off Lid Rack
GREENER_CHOICE
Perform applications accurately and consistently with Barrier Pipette Tips in Lift-off Lid Racks, which seal when exposed to potential contaminants.
Supplier:  Thermo Scientific™ 2065E
 View more versions of this product
Catalog No. 212368
Only null left
Description
Description
Perform applications consistently and accurately by using Thermo Scientific™ ART™ Barrier Pipette Tips in Lift-off Lid Rack, which seal when exposed to potential contaminants, trapping them inside the barrier. ART barrier pipette tips are designed to aid in pipetting for a variety of applications. Extended length, Thermo Scientific™ REACH™ tips provide an extra level of contamination control.
Features:
  • Sterile tips are certified free from RNase, DNase and pyrogen
  • Available in sterile, nonsterile and in various packaging options
  • Manufactured according to the industry's highest quality standard
Designed to aid in pipetteting for a variety of applications:
Extended length to prevent contamination by keeping the pipettete shaft from entering the sample vessel
Gel loading for loading acrylamide or agarose gels
Wide orifice genomic for genomic DNA, fragile cell lines and other viscous materials
Solvent Safe for acids, bases and aggressive organic solvents
Compatible with:
SoftFit™ pipette tips provide precise and consistent pipetteting with multichannel pipettes
Ergonomic pipette tips are designed for Gilson pipetteman™ pipettes and Thermo Scientific™ Finnpipette™ Focus pipettes
Specifications
Chemical Identifiers
Filtered
Not Autoclavable
Lift-off Lid Rack
ART
Sterile
DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free
Universal Fit
Polypropylene
Specifications
ART 100E, Filtered, Sterile, Lift-off Lid Rack
100μL
Sterile
ART 100E
5.07 cm
Lift-off Lid Rack
Universal Fit
Polypropylene
100 μL
Filtered
Not Autoclavable
2 in.
DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free
Micro Point
ART
Certified Quality
Product Suggestions
Product Suggestions