missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ ART™ Barrier Pipette Tips in Bulk Packaging
GREENER_CHOICE

Catalog No. 2123629
Change view
Click to view available options
Volume (Metric):
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
2123629 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 2123629 Supplier Thermo Scientific™ Supplier No. 2180B

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Scientific™ ART™ Barrier Pipette tips seal in contaminants, providing bi-directional protection for samples and pipettes. Fits most pipettes.

ART™ Barrier tips are not the same as traditional filter pipette tips. ART Barrier tips contain a one-of-a-kind, self-sealing barrier that seals in aerosol and liquid contaminants on contact, providing bi-directional protection from contamination for pipettes and samples.

  • Universal Tip, fits most brands of pipettes
  • ART™ tips are certified to be free DNase, RNase, DNA, ATP, Endotoxin, and Pyrogens
  • Convenient bulk packaging for high volume usage

  • Sterile
  • Low Retention
  • Racked with lift-off lid

Specifications

Description ART 5000, Filtered, Sterile, Bulk
Volume (Metric) 5 mL
Filtered Filtered
Sterility Sterile
Color Clear
Autoclavable Not Autoclavable
Model ART 5000
Clean Claims DNase, RNase, DNA, ATP, Bioburden, PCR Inhibitor, Endotoxin, and Pyrogen Free
For Use With (Application) Cell Culture, Pipetting Large volumes of liquids where great accuracy and precision is needed
Format Bulk
Tip Style Macro
Packaging Bag of 250
Quantity 250 tips
Disposable Single-use
For Use With (Equipment) Universal Fit
Product Line ART
Material Virgin Polypropylene
Show More Show Less