Learn More
Thermo Scientific™ ART™ Barrier Low-Retention Pipette Tips
GREENER_CHOICE
Catalog No. 214025e007
Change view
Click to view available options
Volume (Metric):
10 μL
20 μL
100 μL
200 μL
1000 μL
Model:
ART 10
ART 10 REACH
ART 1000
ART 100E
ART 10F
ART 200
ART 20P
Tip Style:
Ergonomic, Low Retention
Extended Length, Low Retention, Micro Point
Low Retention, Micro Point
Fit for Purpose:
Sterile and PCR Ready
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Model Tip Style Fit for Purpose
214025E007 10 μL ART 10 Low Retention, Micro Point Sterile and PCR Ready
21402482 10 μL ART 10 REACH Extended Length, Low Retention, Micro Point Sterile and PCR Ready
21402481 10 μL ART 10F Low Retention, Micro Point Sterile and PCR Ready
21402484 20 μL ART 20P Low Retention, Micro Point Sterile and PCR Ready
21402485 100 μL ART 100E Low Retention, Micro Point -
21402486 200 μL ART 200 Low Retention, Micro Point Sterile and PCR Ready
21402487 1000 μL ART 1000 Ergonomic, Low Retention Sterile and PCR Ready
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 214025E007 Supplier Thermo Scientific™ Supplier No. 213905
Only null left
Eliminate sample retention with these specially designed pipette tips.
Get Thermo Scientific™ ART™ Barrier Low-Retention Pipette Tips, which are specifically designed for eliminating sample retention within the pipette tip.
  • Inner surface is extremely hydrophobic, increasing sample reproducibility and sample accuracy
  • Crystal clear tips allow you to easily see transparent samples
  • Tips seal when exposed to potential contaminants
  • Certified RNase, DNase, ATP, Endotoxin, and Pyrogen Free
Specifications
Description ART 10, Filtered, Sterile, Rack
Volume (Metric) 10 μL
Filtered Filtered
Sterility Sterile
Color Clear
Autoclavable Not Autoclavable
Model ART 10
Clean Claims DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free
Format Rack
Tip Style Low Retention, Micro Point
Packaging Rack, 10 x 96
Product Line ART
Fit for Purpose Sterile and PCR Ready
Show More Show Less