missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ ART™ Barrier Extended Length Pipette Tips
GREENER_CHOICE

Perform pipetting accurately and consistently with Thermo Scientific™ ART™ Extended Length Barrier Pipette Tips, which seal when exposed to potential contaminants.

$466.84 - $659.36

Specifications

Filtered Filtered
Sterility Sterile
Clean Claims DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free
Format Hinged Rack
For Use With (Equipment) Universal Fit
View More Specs

Products 9
Catalog Number Mfr. No. Volume (Metric) Model Tip Style Fit for Purpose Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Model Tip Style Fit for Purpose Price Quantity & Availability  
02682258
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
2140HR
10 μL ART 10 REACH Extended Length Sterile and PCR Ready
Pack of 960 Each for $490.94
Case of 5 Pack for $2,454.70
Only null left
 
02682259
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
214005HR
10 μL ART 10 REACH Low Retention Sterile and PCR Ready
Pack of 960 Each for $548.52
Case of 5 Pack for $2,742.60
Only null left
 
13811138
GREENER_CHOICE
View Documents Promotion Details
N/A 200 μL ART Barrier Universal Fit Sterile and PCR Ready
Pack of 768 Each for $591.57
Case of 5 Pack for $2,957.85
Only null left
 
13811139
GREENER_CHOICE
View Documents Promotion Details
N/A 200 μL ART Barrier Low Retention Sterile and PCR Ready
Pack of 768 Each for $659.36
Case of 5 Pack for $3,296.80
Only null left
 
13811164
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
2079HR
1000 μL ART Barrier Universal Fit Sterile and PCR Ready
Pack of 768 Each for $466.84
Case of 4 Pack for $1,867.36
Only null left
 
13811165
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
207905HR
1000 μL ART Barrier Low Retention Sterile and PCR Ready
Pack of 768 Each for $559.84
Case of 4 Pack for $2,239.36
Only null left
 
13811170
GREENER_CHOICE
View Documents Promotion Details
N/A 1250 μL ART Barrier Extended Length Sterile and PCR Ready
Pack of 768 Each for $554.61
Case of 4 Pack for $2,218.44
Only null left
 
13811171
GREENER_CHOICE
View Documents Promotion Details
N/A 1250 μL ART Barrier Low Retention Sterile and PCR Ready
Pack of 768 Each for $631.61
Case of 4 Pack for $2,526.44
Only null left
 
02682269
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
217905HR
1000 μL ART 1000L Low Retention Sterile and PCR Ready
Pack of 768 Each for $518.35
Case of 4 Pack for $2,073.40
Only null left
 
Description

Description

Unlike standard filter tips, Thermo Scientific™ ART™ Barrier Extended Length Pipette Tips in Hinged Racks seal when exposed to potential contaminants. The pipette tips from the reload inserts fit securely into the environmentally friendly, reusable hinged racks and the covers may be used as hinged or lift-off style. The locking clasp prevents contamination by keeping remaining tips sterile and offers an easy, one-handed operation. ART hinged racks are available in low retention, micropoint, extended length, and other specialty styles.

Features:
  • Stackable hinged racks save space in your lab
  • Sterile tips are DNase, RNase, DNA, ATP, PCR Inhibitor, and Endotoxin Free

Recommended for:

  • Liquid Handling
  • Molecular Biology
  • PCR
  • Biotechnology
Specifications

Specifications

Filtered
DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free
Universal Fit
Sterile and PCR Ready
Sterile
Hinged Rack
ART
Videos
SDS
Documents

Documents