missing translation for 'onlineSavingsMsg'
Learn More

Arsenic Trioxide, Powder, ACS, 99.95-100.05%, Spectrum™ Chemical

Catalog No. p-7165037
Change view
Click to view available options
Quantity:
5 g
25 g
125 g
500 g
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608938 5 g Amber Glass Bottle
18608934 25 g Amber Glass Bottle
18608936 125 g Amber Glass Bottle
18608937 500 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18608938 Supplier Spectrum Chemical Mfg Cor Supplier No. A13405GM
Only null left

Chemical Identifiers

CAS 1327-53-3
Molecular Formula As2O3
Molecular Weight (g/mol) 197.84
MDL Number MFCD00003433
InChI Key IKWTVSLWAPBBKU-UHFFFAOYSA-N
IUPAC Name diarsorosooxidane
SMILES O=[As]O[As]=O

Specifications

CAS Max % 1
Quantity 5 g
Percent Purity 99.95 to 100.05%
Grade ACS
Packaging Amber Glass Bottle
Residue after Ignition 0.0002

CAUTION: For manufacturing or laboratory use only. Not for pesticide use. Read and understand the label and Safety Data Sheet (SDS) prior to use.