Learn More
Arsenic(III) oxide, 99.99% (metals basis)
CAS: 1327-53-3 | As2O3 | 197.84 g/mol
Supplier: Thermo Scientific Chemicals 04037030
Description
used in the manufacturing of such products as glass and enamels. Arsenic trioxide is utilized in various studies on cellular processes, notably in investigations of leukemia and principal material for the production of many arsenic compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 465°C (sublimation) | |
| 312.3°C | |
| White | |
| 250 g | |
| As2O3 | |
| Soluble in dilute acids and alkalies.Insoluble in organic solvents.Slightly soluble in water. | |
| O=[As]O[As]=O | |
| 197.84 | |
| Odorless |
| Arsenic(III) oxide | |
| 1327-53-3 | |
| Powder | |
| 99.99% (Metals basis) | |
| MFCD00003433 | |
| IKWTVSLWAPBBKU-UHFFFAOYSA-N | |
| diarsorosooxidane | |
| 197.84 | |
| 3.74 |
Chemical Identifiers
| 1327-53-3 | |
| 197.84 | |
| IKWTVSLWAPBBKU-UHFFFAOYSA-N | |
| O=[As]O[As]=O |
| As2O3 | |
| MFCD00003433 | |
| diarsorosooxidane |
Safety and Handling
P201-P202-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H300-H314-H335-H350-H371
DOTInformation : Hazard Class: 6.1; Packaging Group: II
EINECSNumber : 215-481-4
RTECSNumber : CG3325000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only