missing translation for 'onlineSavingsMsg'
Learn More

Arsenic(III) oxide, 99.5% (metals basis)

Catalog No. p-7058907
Change view
Click to view available options
Quantity:
25 g
250 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4348814 25 g
AA4348830 250 g
AA43488A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4348814 Supplier Thermo Scientific Chemicals Supplier No. 04348814
Only null left

CAS: 1327-53-3 | As2O3 | 197.84 g/mol

Used in the manufacturing of such products as glass and enamels. Arsenic trioxide is utilized in various studies on cellular processes, notably in investigations of leukemia and principal material for the production of many arsenic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1327-53-3
Molecular Formula As2O3
Molecular Weight (g/mol) 197.84
MDL Number MFCD00003433
InChI Key IKWTVSLWAPBBKU-UHFFFAOYSA-N
Synonym Arsenic trioxide
IUPAC Name diarsorosooxidane
SMILES O=[As]O[As]=O

Specifications

Boiling Point 465°C (sublimation)
Melting Point 312.3°C
Physical Form Powder
Quantity 25 g
UN Number UN1561
Merck Index 14,804
Solubility Information Slightly soluble in cold water; Soluble in boiling water,HCl,alkali hydroxide; Insoluble in alcohol,chloroform,and etherSoluble in dilute acids and alkalies; Insoluble in organic solvents; Slightly soluble in water.
Formula Weight 197.84
Percent Purity 99.5%
Odor Odorless
Density 3.74 g/mL
Assay (metals basis)
Chemical Name or Material Arsenic (III) oxide
Show More Show Less
Hazard Category H300-H314-H335-H350-H371
Hazard Statement GHS H Statement
H300-H350-H314-H318
Fatal if swallowed.
May cause cancer.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ARSENIC TRIOXIDE
EINECSNumber 215-481-4
RTECSNumber CG3325000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only