missing translation for 'onlineSavingsMsg'
Learn More

Arsenazo III Dye Solution (Reagent AR), Reagents

Catalog No. 01336194
Change view
Click to view available options
Quantity:
250 mL
500 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336194 1 L
01336193 250 mL
01336192 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01336194 Supplier Labchem Inc Supplier No. C83029001C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Arsenazo III Dye Solution (Reagent AR), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

  • Arsenazo III Dye Solution is an aqueous indicator dye supplied in a glass amber container.
  • Concentration: 0.05% (W/V)
  • Volume: 500 mL

Specifications

Color Red/Purple
Volume (Metric) 1 L
Packaging Glass Bottle
Quantity 1 L
Sterility Non-sterile
Type Dye Solution