missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Arsenazo III Dye Solution (Reagent AR), Reagents
Description
- Arsenazo III Dye Solution is an aqueous indicator dye supplied in a glass amber container.
- Concentration: 0.05% (W/V)
- Volume: 500 mL
Specifications
Specifications
| Color | Red/Purple |
| Volume (Metric) | 250 mL |
| Quantity | 250 mL |
| Sterility | Non-sterile |
| Type | Indicator Solution |