missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ Arsenal™ 5216 Responder Gear Bag

Catalog No. 19819837
Change view
Click to view available options
Color:
Blue
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19819837 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19819837 Supplier Ergodyne™ Supplier No. 13447

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Arsenal™ 5216 Responder Gear Bag provides convenient and organized access to medical supplies.

  • 600D polyester main material
  • Tarpaulin reinforced bottom with molded feet for extreme durability
  • Top grab handles for rapid response clutch and go
  • Large clamshell side pockets drop down
  • Collapsible internal dividers for customized organization
  • PALS webbing on inside of cover for slotted storage
  • Lock-ready zippers for securing contents
  • Removable shoulder strap for hauling
  • Reflective strips for increased visibility
  • ID window for user identification

Specifications

Color Blue
Material 600D Polyester
Dimensions (L x W x H) 25.5 x 14.5 x 10.5 in. (65 x 37 x 27 cm)
For Use With (Application) Firefighters, EMTs
Quantity 1 Ea.