missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ Arsenal™ 5183 Full Mack Respirator Bag

Catalog No. 19819786
Change view
Click to view available options
Capacity (Metric):
7 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
19819786 7 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19819786 Supplier Ergodyne™ Supplier No. 13183

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Full mack bag makes respiratory equipment easily accessible with hook and loop attachment.

  • Hook and loop allows for easy attachment
  • Cover flap

Specifications

Color Black
Material 420D Nylon
Model 13183
Capacity (Metric) 7 L
For Use With (Application) Construction, Industrial
Quantity 1 Ea.
Dimensions (L x W x H) 9.5 x 4.5 x 10 in. (24 x 11 x 25 cm)