missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ Arsenal™ 5120 Large Wheeled Gear Bag

Catalog No. 191300577
Change view
Click to view available options
Color:
Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
191300577 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 191300577 Supplier Ergodyne™ Supplier No. 13120

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Arsenal™ 5120 Large Wheeled Gear Bag includes a telescoping pull handle and wheels.

  • 1200D polyester main material
  • Molded wheels for safe and easy transport
  • Three removable internal dividers for organization
  • Molded retractable pull handle
  • Padded and adjustable shoulder strap
  • Heavy-gauge nylon stitching and heavy-duty zippers
  • Material backing provides water protection

Specifications

Color Black
Material 1200D Polyester
Dimensions (L x W x H) 32.5 x 14 x 12.5 in. (83 x 35 x 32 cm)
For Use With (Application) Contractors, Trades, Construction, Electricians, General Purpose
Quantity 1 Ea.
Volume (Metric) 93 L