missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Arprinocid 95.0+%, TCI America™
$255.38 - $764.27
Chemical Identifiers
| CAS | 55779-18-5 |
|---|---|
| Molecular Formula | C12H9ClFN5 |
| Molecular Weight (g/mol) | 277.69 |
| MDL Number | MFCD00867210 |
| InChI Key | NAPNOSFRRMHNBJ-UHFFFAOYSA-N |
| Synonym | 9-(2-Chloro-6-fluorobenzyl)-9H-purin-6-amine, Arpocox |
| PubChem CID | 41574 |
| IUPAC Name | 9-[(2-chloro-6-fluorophenyl)methyl]-9H-purin-6-amine |
| SMILES | NC1=C2N=CN(CC3=C(F)C=CC=C3Cl)C2=NC=N1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
A314650MG
|
TCI America
A314650MG |
50 mg |
Each for $255.38
|
|
|||||
|
A3146250MG
|
TCI America
A3146250MG |
250 mg |
Each for $764.27
|
|
|||||
Chemical Identifiers
| 55779-18-5 | |
| 277.69 | |
| NAPNOSFRRMHNBJ-UHFFFAOYSA-N | |
| 41574 | |
| NC1=C2N=CN(CC3=C(F)C=CC=C3Cl)C2=NC=N1 |
| C12H9ClFN5 | |
| MFCD00867210 | |
| 9-(2-Chloro-6-fluorobenzyl)-9H-purin-6-amine, Arpocox | |
| 9-[(2-chloro-6-fluorophenyl)methyl]-9H-purin-6-amine |
Specifications
| 55779-18-5 | |
| C12H9ClFN5 | |
| 50 mg | |
| NAPNOSFRRMHNBJ-UHFFFAOYSA-N | |
| 9-[(2-chloro-6-fluorophenyl)methyl]-9H-purin-6-amine | |
| 41574 | |
| ≥95.0% (HPLC) | |
| Arprinocid |
| 245°C | |
| MFCD00867210 | |
| 9-(2-Chloro-6-fluorobenzyl)-9H-purin-6-amine, Arpocox | |
| NC1=C2N=CN(CC3=C(F)C=CC=C3Cl)C2=NC=N1 | |
| 277.69 | |
| 277.69 | |
| Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator