missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Arprinocid 95.0+%, TCI America™
Specifications
Specifications
| Melting Point | 245°C |
| Quantity | 50 mg |
| Formula Weight | 277.69 |
| Percent Purity | ≥95.0% (HPLC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Arprinocid |
Safety and Handling
| TSCA | No |
| Recommended Storage | Refrigerator |