missing translation for 'onlineSavingsMsg'
Learn More

Arotinolol Hydrochloride 98.0+%, TCI America™

Catalog No. A314550MG
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A314550MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A314550MG Supplier TCI America Supplier No. A314550MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 68377-91-3
Melting Point 236°C
Color White-Yellow
Molecular Formula C15H22ClN3O2S3
Quantity 50 mg
MDL Number MFCD00941421
UN Number 2811
Synonym 5-[2-[[3-(tert-Butylamino)-2-hydroxypropyl]thio]thiazol-4-yl]thiophene-2-carboxamide Hydrochloride
InChI Key XXDAXBZYUXLDRD-UHFFFAOYNA-N
SMILES [H+].[Cl-].CC(C)(C)NCC(O)CSC1=NC(=CS1)C1=CC=C(S1)C(N)=O
IUPAC Name hydrogen 5-(2-{[3-(tert-butylamino)-2-hydroxypropyl]sulfanyl}-1,3-thiazol-4-yl)thiophene-2-carboxamide chloride
Molecular Weight (g/mol) 407.99
PubChem CID 155032
Formula Weight 407.99
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Arotinolol Hydrochloride
Show More Show Less
RTECSNumber XM8301000
TSCA No