Learn More
Sartorius arium™ Water System Cartridge Set
Produce ultrapure water beyond the ASTM Type 1 standard. Sartorius arium™ Water System Cartridge Set is designed for installation in arium™ Pro, Basic or 611 systems. Each consumable kit includes a pre-treatment and polishing cartridge. Kits are specifically formulated for each particular system.
Supplier:  Sartorius H2OAPACK
 View more versions of this product
Catalog No. 14556001
Only null left
Alert:
*Tap water cartridges should be used only after the feed water specifications have been checked and following consultation with the responsible Sartorius™ application specialist.
Description
Description
arium™ water purification systems (Pro, Basic and/or 611 series) are designed to produce the highest purity laboratory water in the industry, with specifications that exceed the ASTM Type 1 standard. The cartridge sets are constructed of the highest quality materials with an advanced design to produce consistent, reproducible results.
  • Exceptional quality of the purification media
    • Optimized to remove organic and inorganic contaminants
    • Highly porous activated carbon combined with efficient ion exchange resins
    • Produces water with the lowest total organic carbon (TOC) content for systems using ultraviolet (UV) technology
  • Specially engineered so that water flows from top to bottom
    • The flow though the ion exchange medium prevents separation of the mixed-bed resin
    • Allows full use of each cartridge with no wasted media
    • Ensures a long cartridge life with unparalleled performance
  • Two-cartridge sets:
    • Pretreatment cartridge
    • Polishing cartridge
  • Specifically formulated for particular arium™ systems:
    • Analytical Kit for VF or UV systems
    • Biological Kit for UF systems
    • Environmental Kit for DI systems
    • Universal Kit for systems with tap water feed*
Specifications
Chemical Identifiers
ASTM type 1
Polypropylene
Activated Carbon and Mixed Bed Ion Exchange Resin
Pretreatment, polishing cartridge
Pretreated
Ion Exchange, Activated Carbon
Standard, cell culture and chromatographic
450 x 230 x 160 mm (WxDxH)
Specifications
ASTM type 1
Pretreated
Ion Exchange, Activated Carbon
Standard, cell culture and chromatographic
Pretreatment, polishing cartridge
450 x 230 x 160 mm (WxDxH)
To 18.2 MΩ x cm ultrapure water in eq: 1.25
Polypropylene
Activated Carbon and Mixed Bed Ion Exchange Resin
arium™ Pro/611 VF or UV Systems
Analytical Kit
Product Suggestions
Product Suggestions