missing translation for 'onlineSavingsMsg'
Learn More

Charcoal, Activated, Norit(R) CA3, Powder, Spectrum™ Chemical

Catalog No. 18609661
Change view
Click to view available options
Quantity:
500 g
2.5 kg
20 kg
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18609661 20 kg Amber Glass Bottle
18609659 500 g Amber Glass Bottle
18609660 2.5 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18609661 Supplier Spectrum Chemical Mfg Cor Supplier No. C119120KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7440-44-0
Molecular Formula C
Molecular Weight (g/mol) 12.01
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N
IUPAC Name carbon
SMILES [C]

Specifications

CAS Max % 1
pH 2.0 to 3.5
Packaging Amber Glass Bottle
Quantity 20 kg
Grade Ungraded
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Charcoal, Activated, Norit(R) CA3, Powder

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.