Learn More
Cole-Parmer™ Essentials Mini Serological Pipettes
An alternative for cramped working spaces, this mini pipette saves you 10.2 cm (4 in.) off standard pipettes!
$290.60 - $471.33
Specifications
Autoclavable Not Autoclavable
Calibrated Yes
Calibration To Deliver
Material Polystyrene
Sterility Sterile
View More Specs
Products 3
Catalog Number Mfr. No. Volume (Metric) Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Price Quantity  
03-395-181
View Documents
Cole-Parmer™ Essentials
2299930
5 mL
Case of 250 Each for $391.40
Only null left
50-121-4572
View Documents
Cole-Parmer™ Essentials
2299931
10 mL
Case of 150 Each for $290.60
Only null left
03-395-183
View Documents
Cole-Parmer™ Essentials
2299932
25 mL
Case of 100 Each for $471.33
Only null left
Description
Description
Easy-to-read graduations from +1 - 2% Color-coded rings for easy identification Polystyrene(PS), gamma sterilized and pyrogen free Unique filter plug stays put and eliminates lint fibers Developed for use in cramped spaces such as laminar hoods, these mini serological pipettes measure 22.4 cm (9') long and fit most serological pipettors. The shorter length reduces distance between sample and pipettor, aiding in more ergonomic pipetting. Gamma sterilized; individually-wrapped with easy peel-apart opening. Dispenser box lets you quickly access pipettes without opening the box top.
Specifications
Specifications
Not Autoclavable
To Deliver
Sterile
Yes
Individually Wrapped
Serological Pipet
Yes
Polystyrene
Fume hoods and cramped spaces
9 in.
22.9 cm
Reusable
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.