missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Essentials HandE-Vac Handheld Aspirating System - Accessories

Catalog No. 10987037
Change view
Click to view available options
Model:
EV501
Type:
Instrument Stand
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Type
10987037 EV501 Instrument Stand
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 10987037 Supplier Cole-Parmer™ Essentials Supplier No. 0439605

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Set of tools for aspiration.

Operates with standard in-house vacuum source or with the M-Vac or E-Vac aspiration systems

Specifications

Compatible Tips E-Vac Only
Description HandE-Vac safety stand
Adjustable Tip Spacing No
Autoclavable Autoclavable
Color Black
Disposable Reusable
Grip Type Ergonomic
Material Plastic
Model EV501
Number of Channels 8
Product Line HandE-Vac
Sterility Non-sterile
Type Instrument Stand
Volume (Metric) 4 L
For Use With (Application) HandE-Vac hand held aspirator
For Use With (Equipment) HandE-Vac hand held aspirator
Show More Show Less