Learn More
Cole-Parmer™ Essentials CELLine Bioreactor Flasks
Ideal for monoclonal antibodies and expressed recombinant cellular protein production
Supplier: Cole-Parmer™ Essentials 2440654
Description
- Constructed with a polystyrene housing and molded silicone gas transfer bottom
- Features a 10,000 MWCO regenerated cellulose semi-permeable membrane and 0.2μm bottom vent membrane barrier
- Gamma irradiation sterilized and pressure tested
These bioreactor flasks are viable in vitro alternatives to using mouse ascites, standard cell culture, or tissue culture products. Bioreactors utilize innovative membrane technology to separate the cell cultivation area from the media chamber. Cells and cellular-secreted products with a MWCO over 10kD are retained between two non-cytotoxic membranes. The upper semi-permeable membrane allows continuous nutrient diffusion and waste elimination. The lower membrane provides direct oxygenation and ensures optimum gas exchange. Separate ports give access to either media or cell cultivation chamber allowing easy change of expended medium without interference of cell growth, function, or gas exchange.
Specifications
| 11.83 oz. | |
| 350 mL | |
| For use with CHO, BHK, HEP2 or any other adherent cell line, for monoclonal Antibody production up to 5mg/mL | |
| Rectangular | |
| Single-use | |
| Threaded | |
| Rectangular |
| 2 x 108 cells / cell compartment | |
| Polystyrene | |
| Bioreactor Flask | |
| Not Autoclavable | |
| Yes | |
| Sterile | |
| Non-treated |