missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Arg-Gly-Asp

For cellular and molecular biology applications

$301.86 - $301.86

Specifications

Purification Method HPLC
Common Name RGD
Content And Storage -30°C to -10°C
Format Powder
For Use With (Application) Fragment from cell-attachment domain of fibronectin; a cell-attachment peptide
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ65468LB0
View Documents
Thermo Scientific Chemicals
J65468LB0
5 mg
Each for $301.86
Only null left
 
Description

Description

Fragment from cell-attachment domain of fibronectin; a cell-attachment peptide

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

HPLC
-30°C to -10°C
Fragment from cell-attachment domain of fibronectin; a cell-attachment peptide
Arg-Gly-Asp
Arginyl-glycyl-aspartic acid
RGD
Powder
346.34
RUO - research use only
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only