missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Arg-Gly-Asp

Catalog No. AAJ65468LB0
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ65468LB0 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ65468LB0 Supplier Thermo Scientific Chemicals Supplier No. J65468LB0

For cellular and molecular biology applications

Fragment from cell-attachment domain of fibronectin; a cell-attachment peptide

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Purification Method HPLC
Common Name RGD
Content And Storage -30°C to -10°C
Format Powder
For Use With (Application) Fragment from cell-attachment domain of fibronectin; a cell-attachment peptide
Molecular Weight (g/mol) 346.34
Product Type Arg-Gly-Asp
Quantity 5 mg
Regulatory Status RUO - research use only
Sequence Arginyl-glycyl-aspartic acid

RUO – Research Use Only