missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Arcturus™ RiboAmp™ HS PLUS Kit
$1938.00 - $7280.00
Specifications
| Type | HS PLUS Kit |
|---|---|
| Content And Storage | cDNA synthesis kit, in vitro transcription (IVT) kit, amplification purification kit (purification reagents and columns). Variable storage conditions |
| For Use With (Application) | Microarray Analysis, Real-Time Quantitative PCR (qPCR) |
| For Use With (Equipment) | ArcturusXT™ LCM Instrument |
| Includes | cDNA Synthesis Kit |
Specifications
| HS PLUS Kit | |
| Microarray Analysis, Real-Time Quantitative PCR (qPCR) | |
| cDNA Synthesis Kit | |
| ARCTURUS, RiboAmp | |
| Any Tissue Type, Cell Cultures, LCM Samples |
| cDNA synthesis kit, in vitro transcription (IVT) kit, amplification purification kit (purification reagents and columns). Variable storage conditions | |
| ArcturusXT™ LCM Instrument | |
| Unlabeled | |
| Not Included | |
| Reverse Transcription, In Vitro Transcription, RNA Amplification |