missing translation for 'onlineSavingsMsg'
Learn More

Arctiin 97.0+%, TCI America™

Catalog No. p-7118226
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
10 mg
50 mg
Catalog No. Quantity
A311810MG 10 mg
A311850MG 50 mg
2 options

Catalog No. A311810MG

Supplier: TCI America A311810MG

Only null left

Chemical Identifiers

CAS 20362-31-6
Molecular Formula C27H34O11
Molecular Weight (g/mol) 534.558
MDL Number MFCD00870630
InChI Key XOJVHLIYNSOZOO-SWOBOCGESA-N
Synonym Arctigenin-4-glucoside, (3R,4R)-4-[(3,4-Dimethoxyphenyl)methyl]-3-[[4-(beta-D-glucopyranosyloxy)-3-methoxyphenyl]methyl]dihydro-2(3H)-furanone
PubChem CID 100528
ChEBI CHEBI:80793
IUPAC Name (3R,4R)-4-[(3,4-dimethoxyphenyl)methyl]-3-[[3-methoxy-4-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyphenyl]methyl]oxolan-2-one
SMILES COC1=C(C=C(C=C1)CC2COC(=O)C2CC3=CC(=C(C=C3)OC4C(C(C(C(O4)CO)O)O)O)OC)OC

Specifications

Melting Point 109°C
Color White-Yellow
Quantity 10 mg
Formula Weight 534.56
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Arctiin
TSCA No
Recommended Storage Freezer