missing translation for 'onlineSavingsMsg'
Learn More

Arctiin 97.0+%, TCI America™

Catalog No. A311810MG
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A311810MG 10 mg
A311850MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A311810MG Supplier TCI America Supplier No. A311810MG

Chemical Identifiers

CAS 20362-31-6
Molecular Formula C27H34O11
Molecular Weight (g/mol) 534.558
MDL Number MFCD00870630
InChI Key XOJVHLIYNSOZOO-SWOBOCGESA-N
Synonym Arctigenin-4-glucoside, (3R,4R)-4-[(3,4-Dimethoxyphenyl)methyl]-3-[[4-(beta-D-glucopyranosyloxy)-3-methoxyphenyl]methyl]dihydro-2(3H)-furanone
PubChem CID 100528
ChEBI CHEBI:80793
IUPAC Name (3R,4R)-4-[(3,4-dimethoxyphenyl)methyl]-3-[[3-methoxy-4-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyphenyl]methyl]oxolan-2-one
SMILES COC1=C(C=C(C=C1)CC2COC(=O)C2CC3=CC(=C(C=C3)OC4C(C(C(C(O4)CO)O)O)O)OC)OC

Specifications

Melting Point 109°C
Color White-Yellow
Quantity 10 mg
Formula Weight 534.56
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Arctiin
TSCA No
Recommended Storage Freezer