missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ (-)-Arctiin

Chemical Identifiers

CAS 20362-31-6
Chemical Name or Material (-)-Arctiin
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC466050010
View Documents
Thermo Scientific™
466050010
1 g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC466052500
View Documents
Thermo Scientific™
466052500
250 mg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

20362-31-6
(-)-Arctiin

Specifications

20362-31-6
1 g
109.0°C to 112.0°C
(-)-Arctiin
Videos
SDS
Documents

Documents

RUO – Research Use Only