Learn More
Thermo Scientific™ Arctic Express™ Dual Storage Systems
Store and ship samples with Arctic Express dual storage systems, which feature hard-shell shipping containers.
Supplier:  Thermo Scientific™ CK50921
 View more versions of this product
Catalog No. 11675901
Only null left
Description
Description
Protect nonhazardous samples during shipment and storage using Thermo Scientific™ Arctic Express™ Storage Systems. Featuring excellent flexibility and safety features, these systems can be used as dry shippers or laboratory cryo vessels.
Features:
  • When filled with liquid nitrogen and placed in the laboratory, unit can be used as a “can and cane” storage system indefinitely with static holding times of up to 125 days.
  • Aluminum vessels come with lockable lid and hold six numbered, color-coded, stainless-steel cans (included), which accommodate canes that hold ampules.
Specifications
Chemical Identifiers
Manual Fill LN2 Dewar
1
Specifications
Manual Fill LN2 Dewar
1
21 shipper/125 lab
2.2 in.
5.6 cm
18.5 L
Arctic Express Dual 19
14.5 in.
36.8 cm
175 x 2mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Accessories