missing translation for 'onlineSavingsMsg'
Learn More

United Scientific™ Archimedes' Principle Apparatus, Large

Catalog No. S15905
Change view
Click to view available options
Product Type:
Density Demonstration Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S15905 Density Demonstration Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S15905 Supplier United Scientific™ Supplier No. BCL002

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Large size enables a group of students to see the demonstration at the same time.

  • This oversized model has a 100 mL displacement capacity for higher accuracy and easier operation. Both the cylinder and the cup have marked divisions, which allow students to repeat the experiment with different volumes.
  • Cylinder is made of white nylon
  • Requires a 200 g spring scale, an overflow can, and a beaker (not included)
  • Includes activity guide

Specifications

For Use With (Application) Spring Scale, Overflow Can, Beaker
Product Type Density Demonstration Kit

Warning: Cancer and reproductive harm