missing translation for 'onlineSavingsMsg'
Learn More

National Safety Apparel™ Enespro™ ArcGuard™ 100 cal Arc Rated Gloves, 18 in. Long

Catalog No. 17001695
Encompass
Change view
Click to view available options
Arc Flash:
100 cal/cm2
65 cal/cm2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Arc Flash
17001695 65 cal/cm2
17001694 100 cal/cm2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17001695 Supplier National Safety Apparel™ Supplier No. G51KDQT14

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Constructed from multiple layers of DuPont™ Nomex and aramid fiber that is inherently flame resistant and will not melt, drip, or ignite.

  • Inherently flame resistant for FR protection that will not wash out
  • Embroidered with arc rating
  • Lined with needle-punched Nomex™ felt for additional flame resistance
  • Wide bell cuff for easy donning and doffing

Specifications

Arc Flash 65 cal/cm2
Certifications/Compliance NFPA 70E, ASTM F1506, OSHA 1910.269, CSA Z462
Applications Industrial
Color Brown
Hand Specific
Material Nomex, Aramid Fiber
Product Type Arc Flash Gloves
Size Universal
For Use With (Application) Arc Flash Hazard Protection
Glove Cuff Color Brown
Glove Cuff Style Open
Series G51KDQ
Length (English) 18 in.
Length (Metric) 45.7 cm
Show More Show Less