missing translation for 'onlineSavingsMsg'
Learn More

National Safety Apparel™ Enespro™ Basic Gear Bag

Catalog No. 17001630
Encompass
Change view
Click to view available options
Size:
Medium
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17001630 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17001630 Supplier National Safety Apparel™ Supplier No. DFDLBAG402

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

  • Adjustable shoulder strap
  • Large front pocket

Specifications

Gender Unisex
For Use With (Application) Utilities, Manufacturing, Electrical Contractors
Material Cotton, Nylon, Polyester
Suspender None
Product Type Gear Bag
Size Medium
Seam Type Stitched
Closure Type Zipper
Color Blue