missing translation for 'onlineSavingsMsg'
Learn More

National Safety Apparel™ Enespro™ ArcGuard™ 65 cal Arc Flash Long Coat/Legging Kit with Standard Hood/Voltage Gloves (11)

Catalog No. p-7220198
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
170011505 Small
170011510 Medium
170011515 Large
170011520 2X-Large
170011525 3X-Large
170011530 4X-Large
170011538 5X-Large
170011539 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 170011505 Supplier National Safety Apparel™ Supplier No. KIT4LC65SM11

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Constructed to be both lightweight and highly durable, this coat and leggings arc flash suit is designed for an arc flash hazard of 65 cal/cm² or less

  • FR Long Coat & Leggings
  • 14 in. Class 2 Rubber Voltage Insulating Gloves
  • 12 in. Leather Glove Protectors
  • Arc Flash Hood Unit with faceshield and hard hat
  • Safety Glasses
  • Faceshield unit bag
  • Glove bag
  • Gear bag

Specifications

Certifications/Compliance CAT 4, NFPA 70E, CSA Z462, ASTM F1506, OSHA 1910.269
Product Type Arc Flash Kit
Gender Unisex
For Use With (Application) Utilities, Manufacturing, Electrical Contractors
Material DuPont™ Nomex™, Kevlar™
Arc Flash Rating 65 cal/cm²
Size Small
Ankle Style Open
Closure Type FR Hook and Loop
Color Brown
Seam Type Stitched
Suspender Adjustable
Show More Show Less