missing translation for 'onlineSavingsMsg'
Learn More

National Safety Apparel™ Enespro™ ArcGuard™ 65 cal Arc Flash Long Coat/Legging Kit with Standard Hood/Voltage Gloves (10)

Catalog No. p-7220197
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
170011504 Small
170011509 Medium
170011514 Large
170011519 2X-Large
170011524 3X-Large
170011529 4X-Large
170011536 5X-Large
170011537 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 170011504 Supplier National Safety Apparel™ Supplier No. KIT4LC65SM10

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Constructed to be both lightweight and highly durable, this coat and leggings arc flash suit is designed for an arc flash hazard of 65 cal/cm² or less

  • Arc flash resistant caramel brown colored coat and leggings, for complete body protection and preventing heat and burns
  • Arc hood unit with faceshield and hard hat, for head and face protection
  • Hood bag
  • Safety glasses
  • Gear bag
  • 14 in. Class 2 rubber insulating gloves, for hand protection against electric shock up to 17,000 VAC
  • 12 in. leather glove protectors, for protecting the rubber gloves from damage
  • Glove bag for glove storage

Specifications

Product Type Arc Flash Kit
Gender Unisex
Certifications/Compliance CAT 4, NFPA 70E, CSA Z462, ASTM F1506, OSHA 1910.269
For Use With (Application) Utilities, Manufacturing, Electrical Contractors
Material DuPont™ Nomex™, Kevlar™
Arc Flash Rating 65 cal/cm²
Seam Type Stitched
Suspender Adjustable
Color Brown
Closure Type FR Hook and Loop
Size Small
Ankle Style Open
Show More Show Less