missing translation for 'onlineSavingsMsg'
Learn More

National Safety Apparel™ Enespro™ 40 cal ArcGuard™ Contractor Arc Flash Kit with Short Coat and Bib Overall (11)

Catalog No. 170011700
Encompass
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
170011700 5X-Large
170011665 Small
170011670 Medium
170011675 Large
170011680 X-Large
170011685 2X-Large
170011690 3X-Large
170011695 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 170011700 Supplier National Safety Apparel™ Supplier No. KIT4SC40EC5X11

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

40 cal arc flash kit with short coat and bib overalls with voltage gloves

  • ArcGuard™ Economy™ Short Coat and Bib Overall
  • 14 in. Class 2 Rubber Insulating Gloves
  • 12 in. Leather Glove Protectors
  • Arc Flash Hood Unit with faceshield & hard hat
  • Safety Glasses
  • Hood bag
  • Glove bag
  • Gear bag

Specifications

Product Type Arc Flash Kit
Certifications/Compliance CAT 4, NFPA 70E, CSA Z462, ASTM F1506, OSHA 1910.269
For Use With (Application) Utilities, Manufacturing, Electrical Contractors
Gender Unisex
Arc Flash Rating 40 cal/cm²
Size 5X-Large
Ankle Style Open
Closure Type FR Hook and Loop
Color Tan
Material Cotton, Nylon
Seam Type Stitched
Suspender Adjustable
Show More Show Less