missing translation for 'onlineSavingsMsg'
Learn More

National Safety Apparel™ Enespro™ 40 cal ArcGuard™ Contractor Arc Flash Kit with Short Coat and Bib Overall (10)

Catalog No. 170011699
Encompass
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
170011699 5X-Large
170011664 Small
170011669 Medium
170011674 Large
170011679 X-Large
170011684 2X-Large
170011689 3X-Large
170011694 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 170011699 Supplier National Safety Apparel™ Supplier No. KIT4SC40EC5X10

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

40 cal arc flash kit with short coat and bib overalls with voltage gloves

  • ArcGuard™ Economy™ Short Coat and Bib Overall
  • 14 in. Class 2 Rubber Insulating Gloves
  • 12 in. Leather Glove Protectors
  • Arc Flash Hood Unit with faceshield & hard hat
  • Safety Glasses
  • Hood bag
  • Glove bag
  • Gear bag

Specifications

Certifications/Compliance CAT 4, NFPA 70E, CSA Z462, ASTM F1506, OSHA 1910.269
For Use With (Application) Utilities, Manufacturing, Electrical Contractors
Product Type Arc Flash Kit
Gender Unisex
Arc Flash Rating 40 cal/cm²
Material Cotton, Nylon
Suspender Adjustable
Ankle Style Open
Closure Type FR Hook and Loop
Color Tan
Seam Type Stitched
Size 5X-Large
Show More Show Less