missing translation for 'onlineSavingsMsg'
Learn More
Learn More
National Safety Apparel™ Enespro™ 25 kA ArcGuard™ Suppression Blanket, 4 ft x 5 ft
Ideal solution for jobs in confined and enclosed spaces where arc flash is a hazard. It is available on its own, or in a convenient kit package which includes the blanket, straps, and carrying bags
$2,712.93 - $3,246.09
Specifications
| Product Type | Fire Blanket |
|---|---|
| Color | Navy |
| Material | Multi-layer Material with Reinforced Holes |
| Certifications/Compliance | NFPA 70 E, ASTM F2676, CSA Z462, OSHA 1910.269, Maximum Arc current Imax = 25 kA |
| Disposable | Limited-use |
Description
- 12 attachment points
- Multi-layer material with reinforced holes for straps
- Provides optimal barrier in enclosed and confined spaces
- Breakopen Threshold Performance (BTP) = 362 kA cycles
- Maximum Arc current Imax = 25 kA
- Made in the USA
Specifications
| Fire Blanket | |
| Multi-layer Material with Reinforced Holes | |
| Limited-use |
| Navy | |
| NFPA 70 E, ASTM F2676, CSA Z462, OSHA 1910.269, Maximum Arc current Imax = 25 kA | |
| Electrical Contractors |