missing translation for 'onlineSavingsMsg'
Learn More

National Safety Apparel™ Enespro™ ArcGuard™ 25 cal Arc Flash Kit with Voltage Gloves (08)

Catalog No. 170011405
Encompass
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
170011405 X-Large
170011374 Small
170011379 Medium
170011384 Large
170011389 2X-Large
170011394 3X-Large
170011399 4X-Large
170011404 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 170011405 Supplier National Safety Apparel™ Supplier No. KIT3SC25XL08

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

ArcGuard™ Arc Flash Kit with Jacket

  • Faceshield Bag
  • Faceshield with hood and hard hat
  • Glove Bag
  • Large Gear Bag
  • Leather Protectors
  • Rubber Voltage Gloves
  • Safety Glasses
  • Bib Overall
  • Short Coat

Specifications

For Use With (Application) Utilities, Manufacturing, Electrical Contractors
Gender Unisex
Product Type Arc Flash Kit
Certifications/Compliance CAT 3, NFPA 70E, ASTM F1506, CSA Z462, OSHA 1910.269
Arc Flash Rating 25 cal/cm²
Suspender Adjustable
Closure Type FR Hook and Loop
Material Cotton, Nylon
Color Khaki
Seam Type Stitched
Size X-Large
Ankle Style Open
Show More Show Less