missing translation for 'onlineSavingsMsg'
Learn More

National Safety Apparel™ Enespro™ ArcGuard™ 20 cal Arc Flash Kit with Voltage Gloves (10)

Catalog No. 170011316
Change view
missing translation for 'orderingAttributeHoverText'
Size:
2X-Large
3X-Large
4X-Large
5X-Large
Large
Medium
Small
X-Large
Catalog No. Size
170011316 2X-Large
170011296 Small
170011301 Medium
170011306 Large
170011311 X-Large
170011321 3X-Large
170011326 4X-Large
170011331 5X-Large
8 options

Catalog No. 170011316

Supplier: National Safety Apparel™ KIT2SC202X10

Only null left

Arc flash kit with PureView™ FR jacket and bib overall

  • PureView™ faceshield technology features a greater range of visibility
  • Premium anti-fog and scratch resistant coating on faceshield
  • FR hook and loop front closure on short coat
  • Stand up collar for extra coverage
  • Rib knit cuffs for comfortable, secure fit
  • Quick disconnect and adjustable shoulder straps on bib overalls
  • Hook and loop at side of bib for easy access
  • Expandable leg opening to fit over boots
  • Made in the USA

Specifications

Gender Unisex
For Use With (Application) Utilities, Manufacturing, Electrical Contractors
Certifications/Compliance CAT 2, NFPA 70E, NFPA 2112, CSA Z462, ASTM F1506, OSHA 1910.269
Product Type Arc Flash Kit
Arc Flash Rating 20 cal/cm²
Closure Type FR Hook and Loop
Size 2X-Large
Suspender Adjustable
Seam Type Stitched
Color Navy
Ankle Style Open
Material Westex™ UltraSoft™
Show More Show Less