missing translation for 'onlineSavingsMsg'
Learn More

National Safety Apparel™ Enespro™ ArcGuard™ 20 cal Coverall Kit with Lift Front Hood and Voltage Gloves (12)

Catalog No. 170011072
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
170011072 5X-Large
170011037 Small
170011042 Medium
170011047 Large
170011052 X-Large
170011057 2X-Large
170011062 3X-Large
170011067 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 170011072 Supplier National Safety Apparel™ Supplier No. KIT2CV20LF5X12

20 cal/cm² Level 2 Arc Flash Kit with an FR Coverall

  • 20 cal FR Coverall
  • 11 in. Class 0 Rubber Voltage Insulating Gloves
  • 10 in. Leather Glove Protectors
  • Hard Hat with Arc Flash Faceshield and Chin Cup
  • Safety Glasses
  • Faceshield unit bag
  • Glove bag
  • Gear bag

Specifications

For Use With (Application) Utilities, Manufacturing, Electrical Contractors
Gender Unisex
Product Type Arc Flash Kit
Material Cotton, Nylon
Certifications/Compliance CAT 2, NFPA 70E, CSA Z462, ASTM F1506, OSHA 1910.269
Arc Flash Rating 20 cal/cm²
Size 5X-Large
Ankle Style Open
Closure Type Zipper
Color Navy
Seam Type Stitched
Suspender None
Show More Show Less