missing translation for 'onlineSavingsMsg'
Learn More

National Safety Apparel™ Enespro™ ArcGuard™ 20 cal Coverall Kit with Lift Front Hood and Voltage Gloves (12)

Catalog No. 170011037
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
170011037 Small
170011042 Medium
170011047 Large
170011052 X-Large
170011057 2X-Large
170011062 3X-Large
170011067 4X-Large
170011072 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 170011037 Supplier National Safety Apparel™ Supplier No. KIT2CV20LFSM12

20 cal/cm² Level 2 Arc Flash Kit with an FR Coverall

  • 20 cal FR Coverall
  • 11 in. Class 0 Rubber Voltage Insulating Gloves
  • 10 in. Leather Glove Protectors
  • Hard Hat with Arc Flash Faceshield and Chin Cup
  • Safety Glasses
  • Faceshield unit bag
  • Glove bag
  • Gear bag

Specifications

Material Cotton, Nylon
Certifications/Compliance CAT 2, NFPA 70E, CSA Z462, ASTM F1506, OSHA 1910.269
For Use With (Application) Utilities, Manufacturing, Electrical Contractors
Gender Unisex
Product Type Arc Flash Kit
Arc Flash Rating 20 cal/cm²
Color Navy
Suspender None
Ankle Style Open
Closure Type Zipper
Seam Type Stitched
Size Small
Show More Show Less