missing translation for 'onlineSavingsMsg'
Learn More

National Safety Apparel™ Enespro™ ArcGuard™ 20 cal Coverall Kit with Lift Front Hood and Voltage Gloves (10)

Catalog No. p-7220251
Change view
missing translation for 'orderingAttributeHoverText'
Size:
2X-Large
3X-Large
4X-Large
5X-Large
Large
Medium
Small
X-Large
Catalog No. Size
170011035 Small
170011040 Medium
170011045 Large
170011050 X-Large
170011055 2X-Large
170011060 3X-Large
170011065 4X-Large
170011070 5X-Large
8 options

Catalog No. 170011035

Supplier: National Safety Apparel™ KIT2CV20LFSM10

Only null left

20 cal/cm² Level 2 Arc Flash Kit with an FR Coverall in UltraSoft

  • 20 cal FR Coverall made from UltraSoft™
  • 11 in. Class 0 Rubber Voltage Insulating Gloves
  • 10 in. Leather Glove Protectors
  • Hard Hat with Arc Flash Faceshield and Chin Cup
  • Safety Glasses
  • Faceshield unit bag
  • Glove bag
  • Gear bag

Specifications

Product Type Arc Flash Kit
Gender Unisex
Certifications/Compliance CAT 2, NFPA 70E, CSA Z462, ASTM F1506, OSHA 1910.269
For Use With (Application) Utilities, Manufacturing, Electrical Contractors
Arc Flash Rating 20 cal/cm²
Material Westex™ UltraSoft™
Color Navy
Seam Type Stitched
Ankle Style Open
Size Small
Closure Type Zipper
Suspender None
Show More Show Less