missing translation for 'onlineSavingsMsg'
Learn More

National Safety Apparel™ Enespro™ ArcGuard™ 12 Cal Long Coat/Legging Kit with Balaclava/Voltage Gloves (12)

Catalog No. 170011208
Encompass
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
170011208 5X-Large
170011173 Small
170011178 Medium
170011183 Large
170011188 X-Large
170011193 2X-Large
170011198 3X-Large
170011203 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 170011208 Supplier National Safety Apparel™ Supplier No. KIT2LC115X12

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ultimate HRC 2 kit without balaclava and gloves

  • Coveralls
  • Hard Hat with ArcGuard™ Faceshield & Chin Cup
  • Safety Glasses
  • Faceshield unit bag
  • Gear bag

Specifications

Product Type Arc Flash Kit
For Use With (Application) Utilities, Manufacturing, Electrical Contractors
Gender Unisex
Material Cotton, Nylon
Certifications/Compliance CAT 2, NFPA 70E, CSA Z462, ASTM F1506, OSHA 1910.269
Arc Flash Rating 12 cal/cm²
Size 5X-Large
Closure Type FR Hook and Loop
Color Navy
Suspender None
Ankle Style Open
Seam Type Stitched
Show More Show Less