missing translation for 'onlineSavingsMsg'
Learn More

National Safety Apparel™ Enespro™ ArcGuard™ 12 Cal Long Coat/Legging Kit with Balaclava/Voltage Gloves (12)

Catalog No. 170011183
Change view
missing translation for 'orderingAttributeHoverText'
Size:
2X-Large
3X-Large
4X-Large
5X-Large
Large
Medium
Small
X-Large
Catalog No. Size
170011183 Large
170011173 Small
170011178 Medium
170011188 X-Large
170011193 2X-Large
170011198 3X-Large
170011203 4X-Large
170011208 5X-Large
8 options

Catalog No. 170011183

Supplier: National Safety Apparel™ KIT2LC11LG12

Only null left

Ultimate HRC 2 kit without balaclava and gloves

  • Coveralls
  • Hard Hat with ArcGuard™ Faceshield & Chin Cup
  • Safety Glasses
  • Faceshield unit bag
  • Gear bag

Specifications

For Use With (Application) Utilities, Manufacturing, Electrical Contractors
Gender Unisex
Product Type Arc Flash Kit
Certifications/Compliance CAT 2, NFPA 70E, CSA Z462, ASTM F1506, OSHA 1910.269
Material Cotton, Nylon
Arc Flash Rating 12 cal/cm²
Color Navy
Closure Type FR Hook and Loop
Size Large
Suspender None
Ankle Style Open
Seam Type Stitched
Show More Show Less