missing translation for 'onlineSavingsMsg'
Learn More

National Safety Apparel™ Enespro™ ArcGuard™ 12 Cal Long Coat/Legging Kit with Balaclava/Voltage Gloves (08)

Catalog No. 170011189
Change view
missing translation for 'orderingAttributeHoverText'
Size:
2X-Large
3X-Large
4X-Large
5X-Large
Large
Medium
Small
X-Large
Catalog No. Size
170011189 2X-Large
170011169 Small
170011174 Medium
170011179 Large
170011184 X-Large
170011194 3X-Large
170011199 4X-Large
170011204 5X-Large
8 options

Catalog No. 170011189

Supplier: National Safety Apparel™ KIT2LC112X08

Only null left

Ultimate HRC 2 kit without balaclava and gloves

  • Coveralls
  • Hard Hat with ArcGuard™ Faceshield & Chin Cup
  • Safety Glasses
  • Faceshield unit bag
  • Gear bag

Specifications

For Use With (Application) Utilities, Manufacturing, Electrical Contractors
Gender Unisex
Product Type Arc Flash Kit
Material Cotton, Nylon
Certifications/Compliance CAT 2, NFPA 70E, CSA Z462, ASTM F1506, OSHA 1910.269
Arc Flash Rating 12 cal/cm²
Ankle Style Open
Color Navy
Seam Type Stitched
Size 2X-Large
Suspender None
Closure Type FR Hook and Loop
Show More Show Less