missing translation for 'onlineSavingsMsg'
Learn More

National Safety Apparel™ Enespro™ ArcGuard™ 12 Cal Long Coat/Legging Kit with Balaclava/Voltage Gloves (08)

Catalog No. 170011184
Change view
missing translation for 'orderingAttributeHoverText'
Size:
2X-Large
3X-Large
4X-Large
5X-Large
Large
Medium
Small
X-Large
Catalog No. Size
170011184 X-Large
170011169 Small
170011174 Medium
170011179 Large
170011189 2X-Large
170011194 3X-Large
170011199 4X-Large
170011204 5X-Large
8 options

Catalog No. 170011184

Supplier: National Safety Apparel™ KIT2LC11XL08

Only null left

Ultimate HRC 2 kit without balaclava and gloves

  • Coveralls
  • Hard Hat with ArcGuard™ Faceshield & Chin Cup
  • Safety Glasses
  • Faceshield unit bag
  • Gear bag

Specifications

For Use With (Application) Utilities, Manufacturing, Electrical Contractors
Certifications/Compliance CAT 2, NFPA 70E, CSA Z462, ASTM F1506, OSHA 1910.269
Material Cotton, Nylon
Gender Unisex
Product Type Arc Flash Kit
Arc Flash Rating 12 cal/cm²
Color Navy
Closure Type FR Hook and Loop
Suspender None
Ankle Style Open
Seam Type Stitched
Size X-Large
Show More Show Less