Learn More
National Safety Apparel™ Arc H2O Jacket FL Orange Segmented Reflective Trim
Supplier: National Safety Apparel™ R30RQCX23X
Specifications
| 3X-Large | |
| Polyurethane-coated FR Cotton | |
| Snap, Zipper | |
| Welded | |
| 30.5 in. |
| Unisex | |
| Fluorescent Orange | |
| Arc Rating: 19 cal/cm² (Fluorescent Yellow); 24 cal/cm² (Fluorescent Orange), CAT 2, NFPA 70 E, ANSI/ISEA 107 Type R Class 3, CSA Z462, CSA Z96 x Calss 2, ASTM F2733, ASTM F1891, OSHA 1910.269 | |
| Hi-vis Jacket |