Learn More
National Safety Apparel™ Arc H2O Jacket FL Orange Segmented Reflective Trim
Catalog No. 170012117
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
170012117 Small
170012118 Medium
170012119 Large
170012120 2X-Large
170012121 X-Large
170012122 3X-Large
170012123 4X-Large
170012124 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 170012117 Supplier National Safety Apparel™ Supplier No. R30RQCX2SM
Specifications
Size Small
Gender Unisex
Material Polyurethane-coated FR Cotton
Color Fluorescent Orange
Closure Type Snap, Zipper
Certifications/Compliance Arc Rating: 9.8 cal/cm² , CAT 2, NFPA 70 E, ANSI/ISEA 107 Type R Class 3, CSA Z462, CSA Z96 x Calss 2, ASTM F2733, ASTM F1891, OSHA 1910.269
Seam Type Welded
Product Type Hi-vis Jacket
Length (English) 30.5 in.