Learn More
National Safety Apparel™ Arc Flash Fan Hood 7.5 oz. Navy Tecgen CC With Quilted Liner
Catalog No. 17001712
Encompass
Change view
Click to view available options
Color: Navy
Catalog No.
17001712 Color: Navy
1 options
Catalog No. 17001712 Supplier National Safety Apparel™ Supplier No. H653NNBQT55HTFAN
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Beekeeper type arc flash hood ideal when working around electrical panels and transformers.
  • Anti-fog faceshield
  • One size fits all with the universal adapter
  • Made in the USA
Specifications
Product Type Arc Flash Hood
Size Medium
Material Modacrylic, Cotton, Para-aramid Fiber
Color Navy
Closure Type None
Seam Type Stitched
Certifications/Compliance Arc Rating: 55 cal/cm², CAT 4, NFPA 70 E, CSA Z462, ASTM F2178, OSHA 1910.269
Includes Arc Flash Hood
Style Hood