missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Arbutin 95.0+%, TCI America™
Specifications
Specifications
| Melting Point | 198°C |
| Color | White |
| Quantity | 5 g |
| Formula Weight | 272.25 |
| Percent Purity | ≥95.0% (HPLC) |
| Grade | Not Available |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Arbutin |
Safety and Handling
| RTECSNumber | CE8863000 |
| TSCA | No |