missing translation for 'onlineSavingsMsg'
Learn More

Arbutin 95.0+%, TCI America™

Supplier:  TCI America A052225G

Quantity 25 g

 View more versions of this product

Catalog No. A052225G


Only null left

Description

Description

Specifications

Chemical Identifiers

497-76-7
272.25
BJRNKVDFDLYUGJ-UHFFFAOYNA-N
440936
2-(hydroxymethyl)-6-(4-hydroxyphenoxy)oxane-3,4,5-triol
C12H16O7
MFCD00016915
arbutin, uvasol, 4-hydroxyphenyl beta-d-glucopyranoside, ursin, beta-arbutin, arbutoside, arbutine, arbutyne, ursi, p-arbutin
CHEBI:18305
OCC1OC(OC2=CC=C(O)C=C2)C(O)C(O)C1O

Specifications

Arbutin
198°C
C12H16O7
MFCD00016915
BJRNKVDFDLYUGJ-UHFFFAOYNA-N
2-(hydroxymethyl)-6-(4-hydroxyphenoxy)oxane-3,4,5-triol
440936
272.25
Crystalline Powder
497-76-7
White
25 g
arbutin, uvasol, 4-hydroxyphenyl beta-d-glucopyranoside, ursin, beta-arbutin, arbutoside, arbutine, arbutyne, ursi, p-arbutin
OCC1OC(OC2=CC=C(O)C=C2)C(O)C(O)C1O
272.25
CHEBI:18305
≥95.0% (HPLC)
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : CE8863000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions