missing translation for 'onlineSavingsMsg'
Learn More

Arbutin, 98+%

CAS: 497-76-7 | C12H16O7 | 272.25 g/mol

Supplier:  Thermo Scientific Chemicals L1494503

 View more versions of this product

Catalog No. AAL1494503


Only null left

Description

Description

Arbutin, a glycosylated hydroquinone, is used in studies on melanin biosynthesis and as an inhibitor to identify, differentiate and characterize tyrosinase(s). Also used in sed as a skin-lightening agent in cosmetic products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Arbutin, a glycosylated hydroquinone, is used in studies on melanin biosynthesis and as an inhibitor to identify, differentiate and characterize tyrosinase(s). Also used in sed as a skin-lightening agent in cosmetic products.

Solubility
Soluble in water (50 mg/ml), methanol and dimethyl sulfoxide.

Notes
Hygroscopic, store away from water/moisture. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Specifications

Arbutin
196°C to 200°C
MFCD00016915
89673
14,773
Soluble in water (50mg/ml),methanol and dimethyl sulfoxide.
−65° (c=2 in Water)
(2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-(4-hydroxyphenoxy)oxane-3,4,5-triol
440936
272.26
497-76-7
C12H16O7
1 g
Air sensitive; Hygroscopic
arbutin, uvasol, 4-hydroxyphenyl beta-d-glucopyranoside, ursin, beta-arbutin, arbutoside, arbutine, arbutyne, ursi, p-arbutin
BJRNKVDFDLYUGJ-UHFFFAOYNA-N
OCC1OC(OC2=CC=C(O)C=C2)C(O)C(O)C1O
272.25
CHEBI:18305
≥98%

Chemical Identifiers

497-76-7
272.25
BJRNKVDFDLYUGJ-UHFFFAOYNA-N
440936
(2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-(4-hydroxyphenoxy)oxane-3,4,5-triol
C12H16O7
MFCD00016915
arbutin, uvasol, 4-hydroxyphenyl beta-d-glucopyranoside, ursin, beta-arbutin, arbutoside, arbutine, arbutyne, ursi, p-arbutin
CHEBI:18305
OCC1OC(OC2=CC=C(O)C=C2)C(O)C(O)C1O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 207-850-3

RTECSNumber : CE8863000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only